Please login to your online account to display your discounted pricing

CD304 Mouse anti-Human, APC-eFluor 780, Clone: TNKUSOHA, eBioscience™

Mouse Monoclonal Antibody

$216.05 - $535.05

Specifications

Antigen CD304
Clone TNKUSOHA
Host Species Murine
Gene Alias BDCA-4, Neuropilin-1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124050

 
life technologies
LS47304941
25 Tests Each for $216.05

501124051

 
life technologies
LS47304942
100 Tests Each for $535.05
Description & Specifications

Specifications

Antigen CD304
Clone TNKUSOHA
Host Species Murine
Gene Alias BDCA-4, Neuropilin-1
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC-eFluor 780
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The TNKUSOHA monoclonal antibody recognizes human CD304, which is also known as BDCA-4 and Neuropilin-1. Like CD303, the CD304 antigen is expressed on human plasmacytoid dendritic cells in normal blood and bone marrow. CD304 can also be detected on T follicular helper cells in inflamed tonsils, as well as on some B-cell acute lymphoblastic leukemias. In addition, CD304 is expressed on numerous non-hematopoietic cell types, including neurons, endothelial, and tumor cells. Plasmacytoid dendritic cells play a key role in the innate immune response against viral pathogens and tumors.