Please login to your online account to display your discounted pricing

CD303a Mouse anti-Human, PerCP-eFluor 710, Clone: 201A, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $477.05

Specifications

Antigen CD303a
Clone 201A
Host Species Murine
Gene Alias BDCA-2, Clec4C
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123963

 
life technologies
LS46981841
25 Tests Each for $187.05

501123964

 
life technologies
LS46981842
100 Tests Each for $477.05
Description & Specifications

Specifications

Antigen CD303a
Clone 201A
Host Species Murine
Gene Alias BDCA-2, Clec4C
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

This 201A monoclonal antibody reacts with human CD303a, which is also known as CLEC4C or Blood Dendritic Cell Antigen 2 (BDCA-2). This type II transmembrane protein is expressed almost exclusively on plasmacytoid dendritic cells, which secrete type I interferon during antiviral and antitumor responses. Studies have demonstrated that CD303a binds asialo-galactosyl-oligosaccharides on invading pathogens, resulting in the activation of signaling pathways that lead to CD303a internalization and downregulation of interferon production.