missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD303a Mouse anti-Human, PE-Cy7, Clone: 201A, eBioscience™

Mouse Monoclonal Antibody

$186.12 - $468.60

Specifications

Antigen CD303a
Clone 201A
Host Species Murine
Gene Alias BDCA-2, Clec4C
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123460 Life Technologies
LS25981841
25 Tests Each for $186.12
501128660 Life Technologies
LS25981842
100 Tests Each for $468.60
Specifications

Description

This 201A monoclonal antibody reacts with human CD303a, which is also known as CLEC4C or Blood Dendritic Cell Antigen 2 (BDCA-2). This type II transmembrane protein is expressed almost exclusively on plasmacytoid dendritic cells, which secrete type I interferon during antiviral and antitumor responses. Studies have demonstrated that CD303a binds asialo-galactosyl-oligosaccharides on invading pathogens, resulting in the activation of signaling pathways that lead to CD303a internalization and downregulation of interferon production.
Description & Specifications

Specifications

CD303a
Murine
Flow Cytometry
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.25μg)/test
201A
BDCA-2, Clec4C
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer