missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD303a Mouse anti-Human, PE-Cyanine7, Clone: 201A, eBioscience™

Mouse Monoclonal Antibody

Manufacturer:  Life Technologies LS25981842

 View more versions of this product

Catalog No. 501128660



Description

Description

Description: This 201A monoclonal antibody reacts with human CD303a, which is also known as CLEC4C or Blood Dendritic Cell Antigen 2 (BDCA-2). This type II transmembrane protein is expressed almost exclusively on plasmacytoid dendritic cells, which secrete type I interferon during antiviral and antitumor responses. Studies have demonstrated that CD303a binds asialo-galactosyl-oligosaccharides on invading pathogens, resulting in the activation of signaling pathways that lead to CD303a internalization and downregulation of interferon production. Applications Reported: This 201A antibody has been reported for use in flow cytometric analysis. Applications Tested: This 201A antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.25 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL cell sample + 100 µL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.

00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. This gene encodes a member of the C-type lectin/C-type lectin-like domain superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may play a role in dendritic cell function. Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications

Specifications

CD303a
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q8WTT0
C-type lectin superfamily member 7, Blood dendritic cell antigen 2 protein, BDCA-2, Dendritic lectin, CD303 antigen, CLEC4C, BDCA2, CLECSF11, CLECSF7, DLEC, HECL, C-type lectin domain family 4 member C
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
201A
PE-Cyanine7
CLEC4C
Liquid
CLEC4C
IgG2a, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
170482
Documents

For Research Use Only.