Please login to your online account to display your discounted pricing

CD303a Mouse anti-Human, APC, Clone: 201A, eBioscience™

Mouse Monoclonal Antibody

$189.95 - $485.75

Specifications

Antigen CD303a
Clone 201A
Host Species Murine
Gene Alias BDCA-2, Clec4C
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123227

 
life technologies
LS17981841
25 Tests Each for $189.95

501123228

 
life technologies
LS17981842
100 Tests Each for $485.75
Description & Specifications

Specifications

Antigen CD303a
Clone 201A
Host Species Murine
Gene Alias BDCA-2, Clec4C
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Allophycocyanin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

This 201A monoclonal antibody reacts with human CD303a, which is also known as CLEC4C or Blood Dendritic Cell Antigen 2 (BDCA-2). This type II transmembrane protein is expressed almost exclusively on plasmacytoid dendritic cells, which secrete type I interferon during antiviral and antitumor responses. Studies have demonstrated that CD303a binds asialo-galactosyl-oligosaccharides on invading pathogens, resulting in the activation of signaling pathways that lead to CD303a internalization and downregulation of interferon production.