missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD301b (MGL2) Rat anti-Mouse, eFluor 660, Clone: 11A10-B7, eBioscience™

Rat Monoclonal Antibody

$170.28 - $407.88

Specifications

Antigen CD301b (MGL2)
Clone 11A10-B7
Host Species Rat
Gene Alias Macrophage galactose-type C-type lectins 2
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124332 Life Technologies
LS50301180
25μg Each for $170.28
501124333 Life Technologies
LS50301182
100μg Each for $407.88
Specifications

Description

The monoclonal antibody 11A10-B7 recognizes mouse CD301b. CD301b is also known as macrophage galactose-type C-type lectin 2 (MGL2). CD301b expression is restricted to dermal dendritic cells, connection tissue, and bone marrow derived dendritic cells. Macrophages also express CD301b at low levels. CD301b contains a single carbohydrate recognition domain that recognizes protein antigens that have a galactose and N-acetylgalactosamine (Gal/GalNAc). Gal/GalNAc protein antigens are efficiently internalized through CD301b and presented to antigen specific CD4+ cells via MHC Class II. CD301b+ dendritic cells are believed to play an important role in TH2 priming.
Description & Specifications

Specifications

CD301b (MGL2)
Rat
Flow Cytometry
eFluor 660
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
11A10-B7
Macrophage galactose-type C-type lectins 2
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer