Please login to your online account to display your discounted pricing

CD301b (MGL2) Rat anti-Mouse, Clone: 11A10-B7, eBioscience™

Rat Monoclonal Antibody

$146.45 - $368.30

Specifications

Antigen CD301b (MGL2)
Clone 11A10-B7
Host Species Rat
Gene Alias Macrophage galactose-type C-type lectins 2
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122668

 
life technologies
LS14301180
25μg Each for $146.45

501122669

 
life technologies
LS14301182
100μg Each for $368.30
Description & Specifications

Specifications

Antigen CD301b (MGL2)
Clone 11A10-B7
Host Species Rat
Gene Alias Macrophage galactose-type C-type lectins 2
Species Reactivity Mouse
Applications Immunohistochemistry (Frozen)
Applications Microscopy
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody 11A10-B7 recognizes mouse CD301b. CD301b is also known as macrophage galactose-type C-type lectin 2 (MGL2). CD301b expression is restricted to dermal dendritic cells, connection tissue, and bone marrow derived dendritic cells. Macrophages also express CD301b at low levels. CD301b contains a single carbohydrate recognition domain that recognizes protein antigens that have a galactose and N-acetylgalactosamine (Gal/GalNAc). Gal/GalNAc protein antigens are efficiently internalized through CD301b and presented to antigen specific CD4+ cells via MHC Class II. CD301b+ dendritic cells are believed to play an important role in TH2 priming.