Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD30 (Reed-Sternberg Cell Marker), Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD30 (Reed-Sternberg Cell Marker), Mouse Monoclonal Antibody.

$237.80 - $1,135.49

Specifications

Antigen CD30 (Reed-Sternberg Cell Marker)
Clone CON6D/ C2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS1915P

 
thermo scientific™ lab vision™
MS1915P
Purified Ab with BSA and Azide 1mL Each for $1,039.78

MS1915P0

 
thermo scientific™ lab vision™
MS1915P0
Purified Ab with BSA and Azide 100µL Each for $237.80

MS1915P1

 
thermo scientific™ lab vision™
MS1915P1
Purified Ab with BSA and Azide 500µL Each for $699.61

MS1915P1ABX

 
thermo scientific™ lab vision™
MS1915P1ABX
Ab without BSA/Azide (Frozen) 100µL Each for $784.92

MS1915PABX

 
thermo scientific™ lab vision™
MS1915PABX
Ab without BSA/Azide (Frozen) 200µL Each for $1,135.49

MS1915R7

 
thermo scientific™ lab vision™
MS1915R7
Ready-to-use for IHC (UVLP) 7mL Each for $667.53
Description & Specifications

Specifications

Antigen CD30 (Reed-Sternberg Cell Marker)
Clone CON6D/ C2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Type Primary
Immunogen Hodgkins lymphoma cell line L428
Isotype IgG2a
Purification Method Hodgkin's lymphoma

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD30 (Reed-Sternberg Cell Marker), Mouse Monoclonal Antibody.

CD30 has been extensively studied as an important cell surface marker on many lymphomas of diverse origin and as an activation molecule of T and B cells. CD30 is a 120kDa single chain transmembrane glycoprotein that is derived from a 90kDa precursor by processing in the golgi complex. CD30 first designated Ki-1, is expressed by mononuclear cells in Hodgkin's lymphoma, Reed Sternberg cells and most Anaplastic Large Cell Lymphomas (ALCL). CD30 is also expressed by embryonal carcinomas.

Host Species: Mouse

Clone: CON6D/C2

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Immunogen: Hodgkin's lymphoma cell line L428

Molecular Weight: 120kDa (mature), 90kDa (precursor)

Positive Control: Hodgkin's lymphoma

Cellular Localization: Membrane and cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO