Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD30 (Reed-Sternberg Cell Marker), Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD30 (Reed-Sternberg Cell Marker), Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1915P1ABX

 View more versions of this product

Catalog No. MS1915P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen CD30 (Reed-Sternberg Cell Marker)
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CON6D/ C2
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Murine
Immunogen Hodgkins lymphoma cell line L428
Isotype IgG2a
Purification Method Hodgkin's lymphoma
Quantity 100μL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD30 (Reed-Sternberg Cell Marker), Mouse Monoclonal Antibody.

CD30 has been extensively studied as an important cell surface marker on many lymphomas of diverse origin and as an activation molecule of T and B cells. CD30 is a 120kDa single chain transmembrane glycoprotein that is derived from a 90kDa precursor by processing in the golgi complex. CD30 first designated Ki-1, is expressed by mononuclear cells in Hodgkin's lymphoma, Reed Sternberg cells and most Anaplastic Large Cell Lymphomas (ALCL). CD30 is also expressed by embryonal carcinomas.

Host Species: Mouse

Clone: CON6D/C2

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Immunogen: Hodgkin's lymphoma cell line L428

Molecular Weight: 120kDa (mature), 90kDa (precursor)

Positive Control: Hodgkin's lymphoma

Cellular Localization: Membrane and cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO