Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD30 (Reed-Sternberg Cell Marker), Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD30 (Reed-Sternberg Cell Marker), Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1915PABX

 View more versions of this product

Catalog No. MS1915PABX

  • / Each
 

Description & Specifications

Specifications

Immunogen Hodgkins lymphoma cell line L428
Clone CON6D/ C2
Isotype IgG2a
Format Concentrated
Cross Reactivity Human
Antigen CD30 (Reed-Sternberg Cell Marker)
Quantity 200µL
Applications Immunohistochemistry (Paraffin)
Regulatory Status RUO
Host Species Mouse
Formulation Ab without BSA/Azide (Frozen)
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled
Purification Method Hodgkin's lymphoma
Research Discipline Cancer and Tumor Biology
Type Primary

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD30 (Reed-Sternberg Cell Marker), Mouse Monoclonal Antibody.

CD30 has been extensively studied as an important cell surface marker on many lymphomas of diverse origin and as an activation molecule of T and B cells. CD30 is a 120kDa single chain transmembrane glycoprotein that is derived from a 90kDa precursor by processing in the golgi complex. CD30 first designated Ki-1, is expressed by mononuclear cells in Hodgkin's lymphoma, Reed Sternberg cells and most Anaplastic Large Cell Lymphomas (ALCL). CD30 is also expressed by embryonal carcinomas.

Host Species: Mouse

Clone: CON6D/C2

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Immunogen: Hodgkin's lymphoma cell line L428

Molecular Weight: 120kDa (mature), 90kDa (precursor)

Positive Control: Hodgkin's lymphoma

Cellular Localization: Membrane and cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO