missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD30 Mouse anti-Human, PE, Clone: Ber-H2, eBioscience™

Mouse Monoclonal Antibody

$157.08 - $302.28

Specifications

Antigen CD30
Clone Ber-H2
Host Species Murine
Gene Alias TNFRSF8, Ki-1
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010317 Life Technologies
LS12030941
25 Tests Each for $157.08
5010318 Life Technologies
LS12030942
100 Tests Each for $302.28
Specifications

Description

The Ber-H2 monoclonal antibody reacts with human CD30, a 120kDa transmembrane glycoprotein of the TNFR superfamily. CD30 is expressed on normal activated cells, T and B lymphocytes but not resting lymphocytes. Expression is also found on some lymphomas and embryonal carcinomas as well as HIV, HTLV and EBV infected lymphocytes. Interaction with its ligand, CD30L, leads to signaling and activation of NK kappa B and subsequently regulates apoptosis.
Description & Specifications

Specifications

CD30
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Ber-H2
TNFRSF8, Ki-1
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.06μg)/test