Please login to your online account to display your discounted pricing

CD30 Mouse anti-Human, FITC, Clone: Ber-H2, eBioscience™

Mouse Monoclonal Antibody

$114.55 - $274.05

Specifications

Antigen CD30
Clone Ber-H2
Host Species Murine
Gene Alias TNFRSF8, Ki-1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122057

 
life technologies
LS11030941
25 Tests Each for $114.55

501122058

 
life technologies
LS11030942
100 Tests Each for $274.05
Description & Specifications

Specifications

Antigen CD30
Clone Ber-H2
Host Species Murine
Gene Alias TNFRSF8, Ki-1
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The Ber-H2 monoclonal antibody reacts with human CD30, a 120kDa transmembrane glycoprotein of the TNFR superfamily. CD30 is expressed on normal activated cells, T and B lymphocytes but not resting lymphocytes. Expression is also found on some lymphomas and embryonal carcinomas as well as HIV, HTLV and EBV infected lymphocytes. Interaction with its ligand, CD30L, leads to signaling and activation of NK kappa B and subsequently regulates apoptosis.