missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD3, eFluor 506, clone: 17A2, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS69003282

 View more versions of this product

Catalog No. 501124918



Description

Description

CD3 complex is crucial in transducing antigen-recognition signals into the cytoplasm of T cells and in regulating the cell surface expression of the TCR complex. T cell activation through the antigen receptor (TCR) involves the cytoplasmic tails of the CD3 subunits CD3 gamma, CD3 delta, CD3 epsilon and CD3 zeta. These CD3 subunits are structurally related members of the immunoglobulins super family encoded by closely linked genes on human chromosome 11. The CD3 components have long cytoplasmic tails that associate with cytoplasmic signal transduction molecules. This association is mediated at least in part by a double tyrosine-based motif present in a single copy in the CD3 subunits. CD3 may play a role in TCR-induced growth arrest, cell survival and proliferation. The CD3 antigen is present on 68-82% of normal peripheral blood lymphocytes, 65-85% of thymocytes and Purkinje cells in the cerebellum. It is never expressed on B or NK cells. Decreased percentages of T lymphocytes may be observed in some autoimmune diseases.

Description: The 17A2 monoclonal antibody reacts with the mouse CD3 complex. CD3 subunits gamma, delta and epsilon are required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. Binding of 17A2 to CD3 initiates the intracellular biochemical pathway resulting in cellular activation and proliferation.Applications Reported: This 17A2 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 17A2 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor™ 506 can be excited with the violet laser line (405 nm) and emits at 506 nm. We recommend using a 510/20 band pass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome.Excitation: 405 nm; Emission: 506 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD3
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P04235, P24161, P22646, P11942
FLJ18683, T3E, TCRE
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
17A2
eFluor 506
Cd3g
Liquid
Cd247, Cd3d, Cd3e, Cd3g
IgG2b, kappa, kappa
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
12500, 12501, 12502, 12503
Documents

For Research Use Only.