Please login to your online account to display your discounted pricing

CD3 Rat anti-Mouse, PerCP-eFluor 710, Clone: 17A2, eBioscience™

Rat Monoclonal Antibody

$158.05 - $456.75

Specifications

Antigen CD3
Clone 17A2
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015891

 
life technologies
LS46003280
25μg Each for $158.05

5015892

 
life technologies
LS46003282
100μg Each for $456.75
Description & Specifications

Specifications

Antigen CD3
Clone 17A2
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 17A2 monoclonal antibody reacts with the mouse CD3 complex. CD3 subunits gamma, delta and epsilon are required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. Binding of 17A2 to CD3 initiates the intracellular biochemical pathway resulting in cellular activation and proliferation.