missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD3 Rat anti-Mouse, Clone: 17A2, eBioscience™

Rat Monoclonal Antibody

$64.68 - $421.08

Specifications

Antigen CD3
Clone 17A2
Host Species Rat
Applications Flow Cytometry,Functional Assay,Immunohistochemistry (Frozen)
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012343 Life Technologies
LS14003281
50μg Each for $64.68
5012344 Life Technologies
LS14003282
100μg Each for $117.48
5012346 Life Technologies
LS14003286
1mg Each for $421.08
501129544 Life Technologies
LS14003285
500μg Each for $249.48
Specifications

Description

The 17A2 monoclonal antibody reacts with the mouse CD3 complex. CD3 subunits gamma, delta and epsilon are required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. Binding of 17A2 to CD3 initiates the intracellular biochemical pathway resulting in cellular activation and proliferation.
Description & Specifications

Specifications

CD3
Rat
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
17A2
Flow Cytometry,Functional Assay,Immunohistochemistry (Frozen)
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL