missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD3 Mouse anti-Rat, APC, Clone: G4.18, eBioscience™

Mouse Monoclonal Antibody

$92.40 - $241.56

Specifications

Antigen CD3
Clone eBioG4.18 (G4.18)
Host Species Murine
Gene Alias T3
Species Reactivity Rat
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123025 Life Technologies
LS17003080
25μg Each for $92.40
501123026 Life Technologies
LS17003082
100μg Each for $241.56
Specifications

Description

The eBioG4.18 monoclonal antibody recognizes the rat CD3 protein. CD3 is a critical component of the T-cell receptor (TCR) and is a marker of the T-cell lineage. CD3 is a complex of several subunits and, upon binding of a TCR ligand, participates in transduction of signals from the TCR to the nucleus which results in a variety of cellular responses including transcription of IL-2 and clonal expansion. \i{in vitro\i}, immobilized G4.18 monoclonal antibody has been demonstrated to activate T cells, while soluble G4.18 inhibited allogeneic mixed-lymphocyte proliferative responses and cell-mediated cytotoxicity to allogeneic target cells. \i{in vivo\i}, G4.18 was demonstrated to induce long-term specific tolerance to an organ allograft.
Description & Specifications

Specifications

CD3
Murine
Rat
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBioG4.18 (G4.18)
T3
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL