missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD3 Mouse anti-Human, PerCP-eFluor 710, Clone: SK7, eBioscience™

Mouse Monoclonal Antibody

$191.40 - $487.20

Specifications

Antigen CD3
Clone SK7
Host Species Murine
Gene Alias Leu-4, T3
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015895 Life Technologies
LS46003641
25 Tests Each for $191.40
5015896 Life Technologies
LS46003642
100 Tests Each for $487.20
Specifications

Description

The SK7 monoclonal antibody reacts with human CD3e, a 20kDa subunit of the TCR complex. Along with the other CD3 subunits γ and δ, the ε chain is required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. The SK7 and UCHT1 monoclonal antibodies cross-block binding, suggesting recognition of overlapping epitopes.

The antibody SK7 recognizes chimpanzee CD3.
Description & Specifications

Specifications

CD3
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
SK7
Leu-4, T3
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.06μg)/test