missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD3 Mouse anti-Human, PerCP-Cy5.5, Clone: SK7, eBioscience™

Mouse Monoclonal Antibody

$443.52 - $443.52

Specifications

Antigen CD3
Clone SK7
Host Species Murine
Gene Alias Leu-4, T3
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015742 Life Technologies
LS45003642
100 Tests Each for $443.52
Specifications

Description

The SK7 monoclonal antibody reacts with human CD3e, a 20kDa subunit of the TCR complex. Along with the other CD3 subunits γ and δ, the ε chain is required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. The SK7 and UCHT1 monoclonal antibodies cross-block binding, suggesting recognition of overlapping epitopes.

The antibody SK7 recognizes chimpanzee CD3.
Description & Specifications

Specifications

CD3
Murine
Flow Cytometry
PerCP-Cy5.5
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.03μg)/test
SK7
Leu-4, T3
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer