Please login to your online account to display your discounted pricing

CD3 Mouse anti-Human, PE, Clone: SK7, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $404.55

Specifications

Antigen CD3
Clone SK7
Host Species Mouse
Gene Alias Leu-4, T3
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50245255

 
life technologies
LS12003641
25 tests Each for $172.55

501129482

 
life technologies
LS12003642
100 tests Each for $404.55
Description & Specifications

Specifications

Antigen CD3
Clone SK7
Host Species Mouse
Gene Alias Leu-4, T3
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The SK7 monoclonal antibody reacts with human and chimpanzee CD3e, a 20kDa subunit of the TCR complex. Along with the other CD3 subunits gamma and delta, the epsilon chain is required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. The SK7 and UCHT1 monoclonal antibodies cross-block binding, suggesting recognition of overlapping epitopes.