Please login to your online account to display your discounted pricing

CD3 Mouse anti-Human, PE, Clone: HIT3a, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $332.05

Specifications

Antigen CD3
Clone HIT3a
Host Species Mouse
Gene Alias Leu-4, T3
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010151

 
life technologies
LS12003941
25 tests Each for $143.55

5010152

 
life technologies
LS12003942
100 tests Each for $332.05
Description & Specifications

Specifications

Antigen CD3
Clone HIT3a
Host Species Mouse
Gene Alias Leu-4, T3
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The HIT3a monoclonal antibody reacts with human CD3e, a 20kDa subunit of the TCR complex. Along with the other CD3 subunits γ and δ, the ε chain is required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. Crosslinking of TCR with HIT3a initiates an intracellular biochemical pathway resulting in cellular activation and proliferation.