Please login to your online account to display your discounted pricing

CD3 Mouse anti-Human, eFluor 450, Clone: UCHT1, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $361.05

Specifications

Antigen CD3
Clone UCHT1
Host Species Mouse
Gene Alias Leu-4, T3
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016230

 
life technologies
LS48003841
25 tests Each for $158.05

5016232

 
life technologies
LS48003880
25μg Each for $158.05

5016233

 
life technologies
LS48003882
100μg Each for $332.05

501128734

 
life technologies
LS48003842
100 tests Each for $361.05
Description & Specifications

Specifications

Antigen CD3
Clone UCHT1
Host Species Mouse
Gene Alias Leu-4, T3
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The UCHT1 monoclonal antibody reacts with human CD3e, a 20kDa subunit of the TCR complex. Along with the other CD3 subunits gamma and delta, the epsilon chain is required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. Crosslinking of TCR via immobilized UCHT1 initiates an intracellular biochemical pathway resulting in cellular activation and proliferation.