Please login to your online account to display your discounted pricing

CD3 Mouse anti-Human, Clone: SK7, Functional Grade, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $477.05

Specifications

Antigen CD3
Clone SK7
Host Species Murine
Gene Alias Leu-4, T3
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014189

 
life technologies
LS16003681
50μg Each for $158.05

5014190

 
life technologies
LS16003685
500μg Each for $477.05

501129519

 
life technologies
LS16003781
50μg Each for $158.05
Description & Specifications

Specifications

Antigen CD3
Clone SK7
Host Species Murine
Gene Alias Leu-4, T3
Species Reactivity Human
Applications Flow Cytometry
Applications Functional Assay
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide

The SK7 monoclonal antibody reacts with human CD3e, a 20kDa subunit of the TCR complex. Along with the other CD3 subunits γ and δ, the ε chain is required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. The SK7 and UCHT1 monoclonal antibodies cross-block binding, suggesting recognition of overlapping epitopes.

The antibody SK7 recognizes chimpanzee CD3.