missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD3 Mouse anti-Human, Clone: HIT3a, Functional Grade, eBioscience™

Mouse Monoclonal Antibody

$143.88 - $434.28

Specifications

Antigen CD3
Clone HIT3a
Host Species Murine
Gene Alias Leu-4, T3
Applications Flow Cytometry,Functional Assay
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014195 Life Technologies
LS16003981
50μg Each for $143.88
5014196 Life Technologies
LS16003985
500μg Each for $434.28
Specifications

Description

The HIT3a monoclonal antibody reacts with human CD3e, a 20kDa subunit of the TCR complex. Along with the other CD3 subunits gamma and delta, the epsilon chain is required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. Crosslinking of TCR with HIT3a initiates an intracellular biochemical pathway resulting in cellular activation and proliferation.
Description & Specifications

Specifications

CD3
Murine
Flow Cytometry,Functional Assay
Unlabeled
Store at 2-8°C.
Monoclonal
HIT3a
Leu-4, T3
RUO
Purified
Primary
aqueous buffer, no sodium azide