missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD3 Mouse anti-Human, Biotin, Clone: OKT3, eBioscience™

Mouse Monoclonal Antibody

$114.55 - $230.55

Specifications

Antigen CD3
Clone OKT3
Host Species Murine
Gene Alias Leu-4, T3
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011380 Life Technologies
LS13003780
25μg Each for $114.55
501129361 Life Technologies
LS13003782
100μg Each for $230.55
Specifications

Description

The OKT3 monoclonal antibody reacts with an epitope on the epsilon-subunit within the human CD3 complex. The OKT3 antibody has been reported to have potent immunosuppressive properties in vivo and has been proven effective in the treatment of renal, heart and liver allograft rejection. The CD3 subunits, gamma, delta, and epsilon chains, are required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. Crosslinking of TCR initiates an intracellular biochemical pathway resulting in cellular activation and proliferation.
Description & Specifications

Specifications

CD3
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
OKT3
Leu-4, T3
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL