missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD3 Mouse anti-Human, APC-eFluor 780, Clone: UCHT1, eBioscience™

Mouse Monoclonal Antibody

$174.24 - $389.40

Specifications

Antigen CD3
Clone UCHT1
Host Species Murine
Gene Alias Leu-4, T3
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016129 Life Technologies
LS47003841
25 Tests Each for $174.24
501129581 Life Technologies
LS47003842
100 Tests Each for $389.40
Specifications

Description

The UCHT1 monoclonal antibody reacts with human CD3e, a 20kDa subunit of the TCR complex. Along with the other CD3 subunits gamma and delta, the epsilon chain is required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. Crosslinking of TCR via immobilized UCHT1 initiates an intracellular biochemical pathway resulting in cellular activation and proliferation.
Description & Specifications

Specifications

CD3
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
UCHT1
Leu-4, T3
Flow Cytometry
APC-eFluor 780
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.5μg)/test