Please login to your online account to display your discounted pricing

CD3 Mouse anti-Human, APC-eFluor 780, Clone: OKT3, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $487.20

Specifications

Antigen CD3
Clone OKT3
Host Species Murine
Gene Alias Leu-4, T3
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123998

 
life technologies
LS47003741
25 Tests Each for $187.05

501123999

 
life technologies
LS47003742
100 Tests Each for $487.20
Description & Specifications

Specifications

Antigen CD3
Clone OKT3
Host Species Murine
Gene Alias Leu-4, T3
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC-eFluor 780
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The OKT3 monoclonal antibody reacts with an epitope on the epsilon-subunit within the human CD3 complex. The OKT3 antibody has been reported to have potent immunosuppressive properties in vivo and has been proven effective in the treatment of renal, heart and liver allograft rejection. The CD3 subunits, gamma, delta, and epsilon chains, are required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. Crosslinking of TCR initiates an intracellular biochemical pathway resulting in cellular activation and proliferation.