Please login to your online account to display your discounted pricing

CD3 Mouse anti-Human, APC, Clone: SK7, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $404.55

Specifications

Antigen CD3
Clone SK7
Host Species Mouse
Gene Alias Leu-4, T3
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014842

 
affymetrix
17003641
25 tests Each for $187.05

501129634

 
affymetrix
17003642
100 tests Each for $404.55
Description & Specifications

Specifications

Antigen CD3
Clone SK7
Host Species Mouse
Gene Alias Leu-4, T3
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The SK7 monoclonal antibody reacts with human CD3e, a 20kDa subunit of the TCR complex. Along with the other CD3 subunits γ and δ, the ε chain is required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. The SK7 and UCHT1 monoclonal antibodies cross-block binding, suggesting recognition of overlapping epitopes.

The antibody SK7 recognizes chimpanzee CD3.