missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD299 (DC-SIGN/L) Mouse anti-Human, APC, Clone: 16E7, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $433.55

Specifications

Antigen CD299 (DC-SIGN/L)
Clone 1.60E+08
Host Species Murine
Gene Alias Clec4M
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123117 Life Technologies
LS17299941
25 Tests Each for $158.05
501123118 Life Technologies
LS17299942
100 Tests Each for $433.55
Specifications

Description

The 16E7 monoclonal antibody recognizes human CD299, which is also known as DC-SIGNR or L-SIGN. A homolog of DC-SIGN, CD299 is an oligomeric type II transmembrane protein expressed on dendritic cells, macrophages in the lung, and endothelial cells in the liver. This C-type lectin binds ICAM-3 to mediate the interaction between dendritic cells and T cells. CD299 also binds the gp120 protein of HIV and the E2 envelope protein of HCV, thereby playing a role in viral infection.
Description & Specifications

Specifications

CD299 (DC-SIGN/L)
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
1.60E+08
Clec4M
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test