Please login to your online account to display your discounted pricing

CD294 (CRTH2) Rat anti-Human, eFluor 660, Clone: BM16, eBioscience™

Rat Monoclonal Antibody

$143.55 - $375.55

Specifications

Antigen CD294 (CRTH2)
Clone BM16
Host Species Rat
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124328

 
life technologies
LS50294941
25 Tests Each for $143.55

501124329

 
life technologies
LS50294942
100 Tests Each for $375.55
Description & Specifications

Specifications

Antigen CD294 (CRTH2)
Clone BM16
Host Species Rat
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

This BM16 monoclonal antibody reacts with human CD294, which is also known as G protein-coupled receptor-44 (GPR-44) or chemoattractant receptor expressed on Th2 cells (CRTH2). CD294 is expressed on Th2 cells, some CD8+ cells, eosinophils, and basophils. Moreover, CD294 binds prostaglandin D2, which promotes leukocyte chemotaxis during allergic responses. CD294 has been reported to mediate prostaglandin D2-mediated suppression of apoptosis of Th2 cells by activating the PI3K pathway.