missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD290 (TLR10) Mouse anti-Human, PE, Clone: 3C10C5, eBioscience™

Mouse Monoclonal Antibody

$170.28 - $421.08

Specifications

Antigen CD290 (TLR10)
Clone 3C10C5
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122301 Life Technologies
LS12290941
25 Tests Each for $170.28
501122302 Life Technologies
LS12290942
100 Tests Each for $421.08
Specifications

Description

This 3C10C5 monoclonal antibody reacts with human CD290, which is also known as Toll-like receptor 10 (TLR10). This type I transmembrane glycoprotein is expressed on B cells, monocytes, regulatory T cells (Tregs), and plasmacytoid dendritic cells. Moreover, CD290 expression is dependent on Foxp3 and NFAT in Tregs. Studies demonstrate that CD290 can self-dimerize, as well as heterodimerize with other members of the TLR family such as TLR1 and TLR2. Ligand binding results in association of the receptor with the adapter molecule MyD88 and subsequent activation of signaling pathways. Although the ligands of CD290 remain undefined, this receptor has been associated with the immune response against microbial and fungal infection and autoimmune diseases such as Crohn’s disease and systemic lupus erythematosous.
Description & Specifications

Specifications

CD290 (TLR10)
Murine
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test
3C10C5
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer