missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD29 Rat anti-Mouse, Clone: KMI6, eBioscience™

Rat Monoclonal Antibody

$91.08 - $302.28

Specifications

Antigen CD29
Clone KMI6
Host Species Rat
Gene Alias Integrin Beta 1, ITGB1, Fibronectin Receptor Beta, VLA-Beta
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012510 Life Technologies
LS14029281
50μg Each for $91.08
5012511 Life Technologies
LS14029282
100μg Each for $143.88
5012512 Life Technologies
LS14029285
500μg Each for $302.28
Specifications

Description

The KMI6 monoclonal antibody reacts with mouse CD29 (integrin β1), a 120-130kDa member of the β integrin family expressed by leukocytes, endothelial, smooth muscle and epithelial cells. CD29 binds non-covalently with the α integrins CD49a-f, to form the VLA-1 through VLA-6 complexes, respectively, and also with CD51. These αβ integrin heterodimers are capable of mediating a variety of cellular responses including adhesion, trafficking, proliferaton and differentiation. All of the integrins which include CD29 bind to extracellular matrix proteins including collagen, laminin, fibronectin and vitronectin, whereas some CD29-containing integrins also interact with cellular receptors including VCAM-1 and MadCAM-1. The KMI6 antibody has been shown to be an activating antibody for β1 integrins, enhancing lymphocyte binding to fibronectin.
Description & Specifications

Specifications

CD29
Rat
Mouse
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
KMI6
Integrin Beta 1, ITGB1, Fibronectin Receptor Beta, VLA-Beta
Flow Cytometry,Functional Assay,Immunohistochemistry (Frozen),Immunoprecipitation,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL