Please login to your online account to display your discounted pricing

CD29 Mouse anti-Human, PE, Clone: TS2/16, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $332.05

Specifications

Antigen CD29
Clone TS2/16
Host Species Mouse
Gene Alias Fibronectin Receptor Beta
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122222

 
life technologies
LS12029941
25 tests Each for $158.05

501122223

 
life technologies
LS12029942
100 tests Each for $332.05
Description & Specifications

Specifications

Antigen CD29
Clone TS2/16
Host Species Mouse
Gene Alias Fibronectin Receptor Beta
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The TS2/16 monoclonal antibody reacts with human CD29, also known as integrin beta 1, an approximately 130kDa single-pass transmembrane glycoprotein. CD29 complexes with one of nine integrin alpha subunits to form the very late antigen (VLA) subfamily of adhesion molecules. Integrin heterodimers containing CD29 are involved in cell-cell and cell-matrix adhesion. CD29 is expressed broadly on lymphocytes and monocytes, with lower levels of expression on granulocytes. The TS2/16 antibody has been found to possess activating activity for beta 1 integrins.