Please login to your online account to display your discounted pricing

CD29 Mouse anti-Human, Clone: TS2/16, eBioscience™

Mouse Monoclonal Antibody

$85.55 - $172.55

Specifications

Antigen CD29
Clone TS2/16
Host Species Mouse
Gene Alias Fibronectin Receptor Beta
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012513

 
life technologies
LS14029980
25μg Each for $85.55

5012514

 
life technologies
LS14029982
100μg Each for $172.55
Description & Specifications

Specifications

Antigen CD29
Clone TS2/16
Host Species Mouse
Gene Alias Fibronectin Receptor Beta
Species Reactivity Human
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunoprecipitation
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The TS2/16 monoclonal antibody reacts with human CD29, also known as integrin beta 1, an approximately 130kDa single-pass transmembrane glycoprotein. CD29 complexes with one of nine integrin alpha subunits to form the very late antigen (VLA) subfamily of adhesion molecules. Integrin heterodimers containing CD29 are involved in cell-cell and cell-matrix adhesion. CD29 is expressed broadly on lymphocytes and monocytes, with lower levels of expression on granulocytes. The TS2/16 antibody has been found to possess activating activity for beta 1 integrins.