Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD29 (Integrin β -1) Ab-3, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD29 (Integrin β-1) Ab-3, Mouse Monoclonal Antibody.

$274.31 - $1,192.92

Specifications

Antigen CD29 (Integrin beta-1) Ab-3
Clone 7F10
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS1089R7

 
thermo scientific™ lab vision™
MS1089R7
7mL Each for $961.94

MS1089S

 
thermo scientific™ lab vision™
MS1089S
1mL Each for $1,192.92

MS1089S0

 
thermo scientific™ lab vision™
MS1089S0
100μL Each for $274.31

MS1089S1

 
thermo scientific™ lab vision™
MS1089S1
500μL Each for $798.45
Description & Specifications

Specifications

Antigen CD29 (Integrin beta-1) Ab-3
Clone 7F10
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Murine
Regulatory Status RUO
Research Discipline Cell Adhesion
Type Primary
Immunogen Recombinant protein encoding the N-terminal region of human CD29
Isotype IgG1

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD29 (Integrin β-1) Ab-3, Mouse Monoclonal Antibody.

CD29, or integrin Beta-1, belongs to the family of cell adhesion receptors. It was initially characterized independently as protein gpIIa appearing on platelets, as the common Beta subunit of the very late activation antigen (VLA), and as a component of various protein complexes binding to extracellular matrix proteins. CD29 is expressed at the cell surface exclusively as part of a heterodimer, in association with one of at least nine different integrin alpha subunits (alpha1, alpha2, alpha3, alpha4, alpha5, alpha6, alpha7, alpha8, and alphav). With the exception of red blood cells and possible weak expression on granulocytes, CD29 is expressed nearly all cell and tissue types.

Host Species: Mouse

Clone: 7F10

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: N-terminal

Immunogen: Recombinant protein encoding the N-terminal region of human CD29

Molecular Weight: 110kDa (nonreducing) and 130 (reducing)

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO