missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
CD29 (Integrin β-1) Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence, and flow cytometry experiments with Epredia™ CD29 (Integrin β-1) Ab-1.
$326.56 - $826.50
Specifications
Antigen | CD29 (Integrin beta-1) Ab-1 |
---|---|
Clone | 4B7R |
Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
CD29 (integrin Beta-1/gpIIa) is a cell adhesion molecule appearing on platelets, as the common Beta subunit of the very late activation antigen (VLA), and as a component of various protein complexes binding to extracellular matrix proteins. CD29 is expressed at the cell surface exclusively as part of a heterodimer, in association with one of at least nine different integrin alpha subunits (alpha1, alpha2, alpha3, alpha4, alpha5, alpha6, alpha7, alpha8, and alphav). With the exception of red blood cells and possible weak expression on granulocytes, CD29 is expressed on nearly all cell and tissue types.Host Species: Mouse
Clone: 4B7R
Isotype: IgG1/κ
Species Reactivity: Human and Pig. Others not tested.
Epitope: Not determined
Immunogen: Human melanoma V+B2 cell line
Molecular Weight: 110kDa (nonreducing) and 130 (reducing)
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Frozen)
- Immunofluorescence
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
CD29 (Integrin beta-1) Ab-1 | |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen) | |
Unconjugated | |
Mouse | |
Cell Adhesion | |
Human melanoma VplusB2 cell line | |
Primary |
4B7R | |
Monoclonal | |
Concentrated | |
RUO | |
Human, Porcine | |
IgG2a κ |