Please login to your online account to display your discounted pricing

CD29 Armenian Hamster anti-Mouse, Rat, PE, Clone: eBioHMb1-1 (HMb1-1), eBioscience™

Armenian Hamster Monoclonal Antibody

$172.55 - $462.55

Specifications

Antigen CD29
Clone eBioHMb1-1 (HMb1-1)
Host Species Armenian Hamster
Gene Alias Fibronectin Receptor Beta
Species Reactivity Mouse, Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010309

 
affymetrix
12029181
50μg Each for $172.55

5010310

 
affymetrix
12029182
100μg Each for $259.55

5010311

 
affymetrix
12029183
200μg Each for $462.55
Description & Specifications

Specifications

Antigen CD29
Clone eBioHMb1-1 (HMb1-1)
Host Species Armenian Hamster
Gene Alias Fibronectin Receptor Beta
Species Reactivity Mouse, Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The eBioHMb1-1 monoclonal antibody reacts with mouse and rat CD29 (integrin beta 1), a 110-120kDa member of the beta integrin family expressed by leukocytes, endothelial, smooth muscle and epithelial cells. CD29 binds non-covalently with the alpha integrins CD49a-f to form the VLA-1 through VLA-6 complexes, as well as with CD51. These alpha-beta integrin heterodimers are capable of mediating a variety of cellular responses including adhesion, trafficking, proliferaton and differentiation. All integrins which include CD29 bind to extracellular matrix proteins including collagen, laminin, fibronectin and vitronectin, whereas some CD29-containing integrins can also interact with cellular receptors such as VCAM-1 and MadCAM-1.