missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD29 Armenian Hamster anti-Mouse, Rat, FITC, Clone: eBioHMb1-1 (HMb1-1), eBioscience™

Armenian Hamster Monoclonal Antibody

$79.20 - $200.64

Specifications

Antigen CD29
Clone eBioHMb1-1 (HMb1-1)
Host Species Armenian Hamster
Gene Alias Fibronectin Receptor Beta
Species Reactivity Mouse, Rat
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122055 Life Technologies
LS11029180
25μg Each for $79.20
501122056 Life Technologies
LS11029182
100μg Each for $200.64
Specifications

Description

The eBioHMb1-1 monoclonal antibody reacts with mouse and rat CD29 (integrin beta 1), a 110-120kDa member of the beta integrin family expressed by leukocytes, endothelial, smooth muscle and epithelial cells. CD29 binds non-covalently with the alpha integrins CD49a-f to form the VLA-1 through VLA-6 complexes, as well as with CD51. These alpha-beta integrin heterodimers are capable of mediating a variety of cellular responses including adhesion, trafficking, proliferaton and differentiation. All integrins which include CD29 bind to extracellular matrix proteins including collagen, laminin, fibronectin and vitronectin, whereas some CD29-containing integrins can also interact with cellular receptors such as VCAM-1 and MadCAM-1.
Description & Specifications

Specifications

CD29
Armenian Hamster
Mouse, Rat
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBioHMb1-1 (HMb1-1)
Fibronectin Receptor Beta
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL