missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD289 (TLR9) Rat anti-Mouse, Clone: M9.D6, eBioscience™

Rat Monoclonal Antibody

$143.55 - $375.55

Specifications

Antigen CD289 (TLR9)
Clone M9.D6
Host Species Rat
Gene Alias TLR-9, toll-like receptor 9
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013797 Life Technologies
LS14909380
25μg Each for $143.55
5013798 Life Technologies
LS14909382
100μg Each for $375.55
Specifications

Description

M9.D6 is generated against a peptide derived from the extracellular portion of mouse TLR9. Predominantly expressed as an intracellular protein, TLR9 is a ∽115-120kDa molecule which mediates response to unmethylated CpG dinucleotides in bacterial DNA. CpG DNA induces a strong T-helper-1-like inflammatory response and the proliferation of TLR9+ B cells. When stimulated with CpG DNA, TLR9-deficient (TLR9-/-) mice lacked splenocyte proliferation, inflammatory cytokine production from macrophages, and dendritic cell maturation, as compared with normal mice. To date, at least ten members of the Toll family have been identified. This family of type I transmembrane proteins is characterized by an extracellular domain with leucine-rich repeats and a cytoplasmic domain with homology to the type I IL-1 receptor. Members of the TLR family are involved in recognition and response to different microbial components including lipoproteins, peptidoglycans, and nucleic acids and play important roles in innate immunity and inflammation. TLR9 is not detected by flow cytometry using this antibody on RBC-lysed mouse splenocytes stained for intracellular TLR9. This may be due to limitations of antigen detection by flow cytometry. Further studies are needed to determine the relationship between mRNA expression and protein detection.
Description & Specifications

Specifications

CD289 (TLR9)
Rat
Mouse
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
M9.D6
TLR-9, toll-like receptor 9
Flow Cytometry (Intracellular Staining),Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL