missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD282 (TLR2) Rat anti-Mouse, PE, Clone: 6C2, eBioscience™

Rat Monoclonal Antibody

$157.08 - $407.88

Specifications

Antigen CD282 (TLR2)
Clone 6C2
Host Species Rat
Gene Alias TLR-2, toll-like receptor 2
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011241 Life Technologies
LS12902180
25μg Each for $157.08
5011242 Life Technologies
LS12902182
100μg Each for $407.88
Specifications

Description

The 6C2 monoclonal antibody reacts with mouse Toll-like receptor 2 (TLR2). Mouse TLR2 is expressed by the myeloid lineage, including macrophage and dendritic cells in splenocytes and the RAW264.7 cell line. To date, at least twelve members of the Toll family have been identified in human and mouse. This family of type I transmembrane proteins is characterized by an extracellular domain with leucine-rich repeats and a cytoplasmic domain with homology to the type I IL-1 receptor. Two of these receptors, TLR2 and TLR4, are pattern recognition receptors and signaling molecules in response to bacterial lipoproteins and have been implicated in innate immunity and inflammation. TLR2 is expressed on the surface of cells and is responsible for distinguishing different pathogens.
Description & Specifications

Specifications

CD282 (TLR2)
Rat
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
6C2
TLR-2, toll-like receptor 2
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer