missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD282 (TLR2) Mouse anti-Mouse, Biotin, Clone: mT2.7, eBioscience™

Mouse Monoclonal Antibody

$143.88 - $368.28

Specifications

Antigen CD282 (TLR2)
Clone mT2.7
Host Species Murine
Gene Alias toll-like receptor 2
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012239 Life Technologies
LS13902280
25μg Each for $143.88
5012240 Life Technologies
LS13902282
100μg Each for $368.28
Specifications

Description

The mT2.7 monoclonal antibody reacts with mouse Toll-like receptor 2 (TLR2). Mouse TLR2 is expressed by the myeloid lineage, including macrophage and dendritic cells in splenocytes and the RAW264.7 cell line. To date, at least twelve members of the Toll family have been identified in human and mouse. This family of type I transmembrane protein is characterized by an extracellular domain with leucine-rich repeats and a cytoplasmic domain with homology to the type I IL-1 receptor. Two of these receptors, TLR2 and TLR4, are pattern recognition receptors and signaling molecules in response to bacterial lipoproteins and have been implicated in innate immunity and inflammation. TLR2 is expressed on the surface of cells and is responsible for distinguishing different pathogens.
Description & Specifications

Specifications

CD282 (TLR2)
Murine
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
mT2.7
toll-like receptor 2
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer