missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD282 (TLR2) Mouse anti-Human, Alexa Fluor 488, Clone: TL2.1, eBioscience™

Mouse Monoclonal Antibody

$170.28 - $381.48

Specifications

Antigen CD282 (TLR2)
Clone TL2.1
Host Species Murine
Gene Alias toll-like receptor 2
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016838 Life Technologies
LS53992241
25 Tests Each for $170.28
5016839 Life Technologies
LS53992242
100 Tests Each for $381.48
Specifications

Description

The TL2.1 monoclonal antibody reacts with human Toll-like receptor 2 (TLR2). To date, at least ten members of the Toll family have been identified in human. This family of type I transmembrane proteins is characterized by an extracellular domain with leucine-rich repeats and a cytoplasmic domain with homology to the type I IL-1 receptor. Two of these receptors, TLR2 and TLR4, are pattern recognition receptors and signaling molecules in response to bacterial lipoproteins and have been implicated in innate immunity and inflammation. TLR2 is expressed by peripheral blood monocytes and is responsible for distinguishing different pathogens. TL2.1, a blocking antibody, has been used to study the role of TLR-2 as a pattern recognition receptor in microbial lipoprotein/lipopeptide induced cytokine production from human peripheral blood mononuclear cells. TL2.1 has been reported to immunoprecipitate human TLR2 (∽90 kDa) from PBMC and HMEC.
Description & Specifications

Specifications

CD282 (TLR2)
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
TL2.1
toll-like receptor 2
Flow Cytometry
Alexa Fluor 488
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (1.0μg)/test