Please login to your online account to display your discounted pricing

CD281 (TLR1) Rat anti-Mouse, eFluor 660, Clone: eBioTR23 (TR23), eBioscience™

Rat Monoclonal Antibody

$172.55 - $433.55

Specifications

Antigen CD281 (TLR1)
Clone eBioTR23 (TR23)
Host Species Rat
Gene Alias Toll-like Receptor 1
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124479

 
life technologies
LS50901180
25μg Each for $172.55

501124480

 
life technologies
LS50901182
100μg Each for $433.55
Description & Specifications

Specifications

Antigen CD281 (TLR1)
Clone eBioTR23 (TR23)
Host Species Rat
Gene Alias Toll-like Receptor 1
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The eBioTR23 monoclonal antibody reacts with mouse Toll-like receptor 1 (TLR1, CD281). TLR1 is an approximately 98kDa protein belonging to the Toll-like receptor family, and is closely-related to mouse TLR6 in both structure and function. Through the formation of a complex with TLR2, TLR1 is able to bind bacterially-derived triacyl lipopeptides, as well as soluble factors released from Neisseria meningitidis. TLR1 is expressed by macrophages and macrophages in TLR1-deficient mice display impaired production of inflammatory cytokines upon stimulation with triacyl lipopeptides and lipoproteins from mycobacteria.