Please login to your online account to display your discounted pricing

CD28 Mouse anti-Rat, PE, Clone: JJ319, eBioscience™

Mouse Monoclonal Antibody

$129.05 - $404.55

Specifications

Antigen CD28
Clone JJ319
Host Species Mouse
Species Reactivity Rat
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010301

 
affymetrix
12028081
50μg Each for $129.05

5010302

 
affymetrix
12028083
200μg Each for $404.55
Description & Specifications

Specifications

Antigen CD28
Clone JJ319
Host Species Mouse
Species Reactivity Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The JJ319 monoclonal antibody reacts with the rat CD28 molecule, expressed by thymocytes, mature T cells, and a subset of NK cells. CD28 is a ligand for CD80 (B7-1) and CD86 (B7-2) and is a potent costimulator of T cells. Signaling through CD28 augments IL-2 and IL-2 receptor expression as well as cytotoxicity of CD3-activated T cells. No reactivity of JJ319 is observed on mouse T cells.