Please login to your online account to display your discounted pricing

CD28 Mouse anti-Human, PE-eFluor 610, Clone: CD28.2, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $433.55

Specifications

Antigen CD28
Clone CD28.2
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124751

 
life technologies
LS61028941
25 tests Each for $172.55

501124752

 
life technologies
LS61028942
100 tests Each for $433.55
Description & Specifications

Specifications

Antigen CD28
Clone CD28.2
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-eFluor 610
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The CD28.2 monoclonal antibody reacts with the human CD28 molecule, a 44kDa homodimer expressed by thymocytes, mature T cells and plasma cells. CD28 is a ligand for CD80 (B7-1) and CD86 (B7-2) and is a potent co-stimulator of T cells. Signaling through CD28 augments IL-2 and IL-2 receptor expression as well as cytotoxicity of CD3-activated T cells.