missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD28 Mouse anti-Human, Clone: CD28.6, Functional Grade, eBioscience™

Mouse Monoclonal Antibody, Functional Grade

$143.88 - $434.28

Specifications

Antigen CD28
Clone CD28.6
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry,Functional Assay,Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014276 Life Technologies
LS16028881
50μg Each for $143.88
501129408 Life Technologies
LS16028885
500μg Each for $434.28
Specifications

Description

The CD28.6 monoclonal antibody reacts with the human CD28 molecule, a 44kDa homodimer expressed by thymocytes, mature T cells and plasma cells. CD28 is a ligand for CD80 (B7-1) and CD86 (B7-2) and is a potent costimulator of T cells. Signaling through CD28 augments IL-2 and IL-2 receptor expression as well as cytotoxicity of CD3-activated T cells. The CD28.6 antibody has been reported for use in immunoblotting (WB) and neutralization studies and costimulation in the production of IL-2.
Description & Specifications

Specifications

CD28
Murine
Flow Cytometry,Functional Assay,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
1mg/mL
CD28.6
Human
RUO
Purified
Primary
aqueous buffer, no sodium azide