Please login to your online account to display your discounted pricing

CD28 Mouse anti-Human, Clone: CD28.2, Functional Grade, eBioscience™

Mouse Monoclonal Antibody, Functional Grade

$158.05 - $477.05

Specifications

Antigen CD28
Clone CD28.2
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014278

 
life technologies
LS16028981
50μg Each for $158.05

501129356

 
life technologies
LS16028985
500μg Each for $477.05
Description & Specifications

Specifications

Antigen CD28
Clone CD28.2
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
Applications Functional Assay
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

The CD28.2 monoclonal antibody reacts with the human CD28 molecule, a 44kDa homodimer expressed by thymocytes, mature T cells and plasma cells. CD28 is a ligand for CD80 (B7-1) and CD86 (B7-2) and is a potent co-stimulator of T cells. Signaling through CD28 augments IL-2 and IL-2 receptor expression as well as cytotoxicity of CD3-activated T cells.