Please login to your online account to display your discounted pricing

CD28 Mouse anti-Human, Clone: CD28.2, eBioscience™

Mouse Monoclonal Antibody

$71.05 - $143.55

Specifications

Antigen CD28
Clone CD28.2
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012505

 
life technologies
LS14028980
25μg Each for $71.05

5012506

 
life technologies
LS14028982
100μg Each for $143.55
Description & Specifications

Specifications

Antigen CD28
Clone CD28.2
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunoprecipitation
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The CD28.2 monoclonal antibody reacts with the human CD28 molecule, a 44kDa homodimer expressed by thymocytes, mature T cells and plasma cells. CD28 is a ligand for CD80 (B7-1) and CD86 (B7-2) and is a potent co-stimulator of T cells. Signaling through CD28 augments IL-2 and IL-2 receptor expression as well as cytotoxicity of CD3-activated T cells.