missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD28 Mouse anti-Canine, APC, Clone: 5B8, eBioscience™

Mouse Monoclonal Antibody

Manufacturer:  Life Technologies LS17028242

 View more versions of this product

Catalog No. 501129239



Description

Description

The 5B8 monoclonal antibody reacts with canine CD28, which is expressed by thymocytes, a majority of T cells, and a subset of NK cells. CD28 is a ligand for CD80 (B7-1) and CD86 (B7-2) and is a potent costimulator of T cells. Signaling through CD28 augments IL-2 and IL-2 receptor expression as well as cytotoxicity of CD3-activated T cells.

The 5B8 antibody has been reported to have agonist activity. The 5B8 and 1C6 antibodies recognize different epitopes of canine CD28.
Specifications

Specifications

CD28
5B8
APC
Conjugated
IgG1, kappa
RUO
Primary
Canine
Flow Cytometry
5μL (0.25μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Documents

For Research Use Only.