missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD28 Golden Syrian Hamster anti-Mouse, PE-Cy7, Clone: 37.51, eBioscience™

Golden Syrian Hamster Monoclonal Antibody

$236.28 - $394.68

Specifications

Antigen CD28
Clone 37.51
Host Species Golden Syrian Hamster
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015491 Life Technologies
LS25028181
50μg Each for $236.28
5015492 Life Technologies
LS25028182
100μg Each for $394.68
Specifications

Description

The 37.51 monoclonal antibody reacts with the mouse CD28 molecule, a 45kDa homodimer expressed by thymocytes, mature T cells and NK cells. CD28 is a ligand for CD80 (B7-1) and CD86 (B7-2) and is a potent costimulator of T cells. Signaling through CD28 augments IL-2 and IL-2 receptor expression as well as cytotoxicity of CD3-activated T cells.
Description & Specifications

Specifications

CD28
Golden Syrian Hamster
Flow Cytometry
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL
37.51
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer