Please login to your online account to display your discounted pricing

CD279 (PD-1) Mouse anti-Human, PerCP-eFluor 710, Clone: MIH4, eBioscience™

Mouse Monoclonal Antibody

$201.55 - $375.55

Specifications

Antigen CD279 (PD-1)
Clone MIH4
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123985

 
life technologies
LS46996941
25 tests Each for $201.55

501123986

 
life technologies
LS46996942
100 tests Each for $375.55
Description & Specifications

Specifications

Antigen CD279 (PD-1)
Clone MIH4
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (1.0μg)/test

The MIH4 monoclonal antibody reacts with the human PD-1 (programmed death-1), a 55kDa member of the immunoglobulin superfamily. PD-1 contains the immunoreceptor tyrosine-based inhibitory motif (ITIM) and plays a key role in peripheral tolerance and autoimmune disease. PD-1 is expressed predominantly on activated T and B lymphocytes. Two novel members of the B7 family have been identified as the PD-1 ligands, PD-L1 (B7-H1) and PD-L2 (B7-DC). Evidence reported to date suggests overlapping functions for these two PD-1 ligands and their constitutive expression on some normal tissues and upregulation on activated antigen-presenting cells. The MIH4 antibody recognizes a different epitope than antibody clones J105.